Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afmid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111366.1 | NP_001104836.1 | 5 - 6 | 417 | 62 | |
XM_006247823.2 | XP_006247885.1 | 5 - 6 | 433 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |