Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017377.1 | NP_001017377.1 | 1 - 2 | 484 | 64 | |
GenBank mRNA | BC092635.1 | - | 1 - 2 | 484 | 7315721 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |