Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047842.1 | NP_001041307.1 | 10 - 11 | 1410 | 60 | |
XM_006246469.2 | XP_006246531.1 | 7 - 8 | 1193 | 60 | ||
GenBank mRNA | AY383687.1 | - | 10 - 11 | 1410 | 60 | |
BC099219.1 | - | 3 - 4 | 424 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |