Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mslnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767615.1 | XP_008765837.1 | 7 - 8 | 2811 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |