Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007607.1 | NP_001007608.1 | 7 - 8 | 1379 | 67 | |
XM_006245942.2 | XP_006246004.1 | 7 - 8 | 1458 | 67 | ||
XM_008767559.1 | XP_008765781.1 | 7 - 8 | 1398 | 67 | ||
XM_008767560.1 | XP_008765782.1 | 7 - 8 | 1425 | 67 | ||
GenBank mRNA | BC081875.1 | - | 7 - 8 | 1379 | 7107681 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |