Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191825.1 | NP_001178754.1 | 5 - 6 | 1177 | 73 | |
XM_006247353.2 | XP_006247415.1 | 6 - 7 | 1403 | 73 | ||
XM_008768080.1 | XP_008766302.1 | 6 - 7 | 1404 | 73 | ||
XM_008768081.1 | XP_008766303.1 | 6 - 7 | 1442 | 73 | ||
XM_008768082.1 | XP_008766304.1 | 6 - 7 | 3087 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |