Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109209.2 | NP_001102679.2 | 10 - 11 | 1404 | 66 | |
GenBank mRNA | X60660.1 | - | 8 - 9 | 963 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |