Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761807.1 | XP_008760029.1 | 9 - 10 | 1600 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Nuclear Receptor Rattus norvegicus Probe spans exons |