Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106937.1 | NP_001100407.1 | 8 - 9 | 989 | 95 | |
XM_006257159.2 | XP_006257221.1 | 9 - 10 | 1258 | 95 | ||
XM_006257160.2 | XP_006257222.1 | 9 - 10 | 1108 | 95 | ||
XM_006257161.2 | XP_006257223.1 | 9 - 10 | 1255 | 95 | ||
XM_006257162.2 | XP_006257224.1 | 8 - 9 | 1298 | 95 | ||
XM_008773339.1 | XP_008771561.1 | 8 - 9 | 973 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |