Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025288.1 | NP_001020459.1 | 4 - 5 | 471 | 62 | |
GenBank mRNA | BC097356.1 | - | 4 - 5 | 471 | 7442140 | 62 |
Gene Symbol: LOC100909986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003752209.3 | XP_003752257.1 | 5 - 6 | 716 | 62 | |
XM_006257731.2 | XP_006257793.1 | 5 - 6 | 716 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |