Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019354.3 | NP_062227.2 | 4 - 5 | 714 | 135 | |
GenBank mRNA | AB005143.1 | - | 2 - 3 | 341 | 135 | |
AB006613.1 | - | 2 - 3 | 341 | 135 | ||
AB010743.1 | - | 4 - 5 | 135 | |||
AF039033.1 | - | 2 - 3 | 341 | 135 | ||
BC062230.1 | - | 4 - 5 | 6920447 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |