Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014244.1 | NP_001014266.1 | 6 - 7 | 631 | 150 | |
XM_008759774.1 | XP_008757996.1 | 4 - 5 | 1769 | 150 | ||
XM_008759775.1 | XP_008757997.1 | 5 - 6 | 1672 | 150 | ||
XM_008759776.1 | XP_008757998.1 | 4 - 5 | 1769 | 150 | ||
XM_008759777.1 | XP_008757999.1 | 4 - 5 | 1769 | 150 | ||
XM_008759778.1 | XP_008758000.1 | 5 - 6 | 1087 | 150 | ||
XM_008759779.1 | XP_008758001.1 | 5 - 6 | 1244 | 150 | ||
XM_008759780.1 | XP_008758002.1 | 3 - 4 | 273 | 150 | ||
XM_008759781.1 | XP_008758003.1 | 6 - 7 | 568 | 150 | ||
GenBank mRNA | BC079235.1 | - | 6 - 7 | 631 | 7132617 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |