Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044265.1 | NP_001037730.1 | 2 - 3 | 588 | 64 | |
GenBank mRNA | AY325154.1 | - | 500 | 64 | ||
BC097353.1 | - | 2 - 3 | 588 | 7387686 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |