Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001069558.6 | XP_001069558.2 | 6 - 7 | 1091 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Recommended Assay Rattus norvegicus Probe spans exons |