Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106425.1 | NP_001099895.1 | 4 - 5 | 676 | 137 | |
XM_008760864.1 | XP_008759086.1 | 4 - 5 | 685 | 137 | ||
GenBank mRNA | BC158721.1 | - | 4 - 5 | 667 | 9090461 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |