Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107159.1 | NP_001100629.1 | 6 - 7 | 983 | 67 | |
XM_006240758.2 | XP_006240820.1 | 6 - 7 | 903 | 67 | ||
GenBank mRNA | FJ799755.1 | - | 6 - 7 | 983 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |