Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106756.1 | NP_001100226.1 | 1 - 2 | 497 | 99 | |
GenBank mRNA | FQ211881.1 | - | 1 - 2 | 212 | 99 | |
FQ217020.1 | - | 1 - 2 | 213 | 99 | ||
FQ217041.1 | - | 1 - 2 | 211 | 99 | ||
FQ223793.1 | - | 1 - 2 | 243 | 99 | ||
FQ224251.1 | - | 1 - 2 | 212 | 99 | ||
FQ224299.1 | - | 1 - 2 | 213 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Inventoried Probe spans exons |