Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107328.2 | NP_001100798.1 | 2 - 3 | 426 | 104 | |
GenBank mRNA | DQ011669.1 | - | 2 - 3 | 426 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |