Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rftn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135011.1 | NP_001128483.1 | 6 - 7 | 1189 | 59 | |
XM_008766876.1 | XP_008765098.1 | 6 - 7 | 1753 | 59 | ||
GenBank mRNA | FQ232229.1 | - | 2 - 3 | 555 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |