Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013055.1 | NP_001013073.1 | 4 - 5 | 501 | 132 | |
XM_006253790.1 | XP_006253852.1 | 4 - 5 | 463 | 132 | ||
GenBank mRNA | BC091191.1 | - | 4 - 5 | 501 | 7368273 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |