Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108952.1 | NP_001102422.1 | 6 - 7 | 517 | 71 | |
GenBank mRNA | FQ227372.1 | - | 6 - 7 | 509 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |