Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107176.1 | NP_001100646.1 | 1 - 2 | 256 | 62 | |
XM_006249871.2 | XP_006249933.1 | 2 - 3 | 1103 | 62 | ||
XM_006249872.2 | XP_006249934.1 | 2 - 3 | 1662 | 62 | ||
XM_008769554.1 | XP_008767776.1 | 1 - 2 | 1489 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |