Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100984.1 | NP_001094454.1 | 7 - 8 | 649 | 109 | |
XM_006250013.2 | XP_006250075.1 | 6 - 7 | 868 | 109 | ||
XM_008769664.1 | XP_008767886.1 | 4 - 5 | 640 | 109 | ||
GenBank mRNA | JN864041.1 | - | 5 - 6 | 673 | 109 | |
JN864042.1 | - | 5 - 6 | 673 | 109 | ||
JN864045.1 | - | 5 - 6 | 667 | 109 | ||
JN864046.1 | - | 4 - 5 | 559 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |