Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scyl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191828.1 | NP_001178757.1 | 2 - 3 | 533 | 63 | |
XM_008769660.1 | XP_008767882.1 | 1 - 2 | 1359 | 63 | ||
XM_008769661.1 | XP_008767883.1 | 1 - 2 | 1359 | 63 | ||
XM_008769662.1 | XP_008767884.1 | 2 - 3 | 417 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Rattus norvegicus Probe spans exons |