Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025752.2 | NP_001020923.2 | 1 - 2 | 290 | 66 | |
GenBank mRNA | BC098838.1 | - | 1 - 2 | 290 | 7378980 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |