Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025752.2 | NP_001020923.2 | 5 - 6 | 767 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |