Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106327.1 | NP_001099797.1 | 4 - 5 | 808 | 66 | |
XM_006230750.2 | XP_006230812.1 | 4 - 5 | 884 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |