Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024242.1 | NP_001019413.1 | 3 - 4 | 284 | 77 | |
GenBank mRNA | BC093401.1 | - | 3 - 4 | 284 | 7389584 | 77 |
FQ227585.1 | - | 1 - 2 | 83 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |