Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033901.1 | NP_001029073.1 | 5 - 6 | 1178 | 76 | |
GenBank mRNA | BC104695.1 | - | 5 - 6 | 1178 | 7111833 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |