Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013887.1 | NP_001013909.1 | 4 - 5 | 475 | 84 | |
XM_006254546.2 | XP_006254608.1 | 3 - 4 | 454 | 84 | ||
GenBank mRNA | BC079289.1 | - | 4 - 5 | 475 | 7134364 | 84 |
FQ214241.1 | - | 3 - 4 | 328 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |