Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007721.1 | NP_001007722.1 | 1 - 2 | 84 | 62 | |
GenBank mRNA | BC081865.1 | - | 1 - 2 | 84 | 7096470 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |