Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eri3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238735.2 | XP_006238797.1 | 1 - 2 | 358 | 74 | |
XM_006238736.2 | XP_006238798.1 | 1 - 2 | 358 | 74 | ||
XM_006238739.1 | XP_006238801.1 | 1 - 2 | 358 | 74 | ||
XM_233435.6 | XP_233435.5 | 1 - 2 | 358 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |