Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109506.1 | NP_001102976.1 | 3 - 4 | 280 | 89 | |
XM_006255302.2 | XP_006255364.1 | 3 - 4 | 527 | 89 | ||
XM_006255303.2 | XP_006255365.1 | 3 - 4 | 326 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |