Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107164.1 | NP_001100634.1 | 2 - 3 | 1000 | 69 | |
GenBank mRNA | BC158887.1 | - | 2 - 3 | 930 | 69 | |
FQ232766.1 | - | 2 - 3 | 961 | 69 | ||
FQ235088.1 | - | 3 - 4 | 812 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |