Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005540.1 | NP_001005540.1 | 5 - 6 | 925 | 76 | |
XM_008761380.1 | XP_008759602.1 | 6 - 7 | 1423 | 76 | ||
XM_008761381.1 | XP_008759603.1 | 6 - 7 | 1423 | 76 | ||
GenBank mRNA | BC082053.1 | - | 5 - 6 | 925 | 7132147 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons |