Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106449.2 | NP_001099919.1 | 4 - 5 | 566 | 69 | |
NM_001270650.1 | NP_001257579.1 | 5 - 6 | 640 | 69 | ||
NM_001270651.1 | NP_001257580.1 | 5 - 6 | 613 | 69 | ||
NM_001270652.1 | NP_001257581.1 | 4 - 5 | 543 | 69 | ||
NM_001270653.1 | NP_001257582.1 | 4 - 5 | 570 | 69 | ||
XM_006232855.2 | XP_006232917.1 | 5 - 6 | 635 | 69 | ||
XM_008761282.1 | XP_008759504.1 | 4 - 5 | 601 | 69 | ||
GenBank mRNA | FN800735.1 | - | 4 - 5 | 524 | 69 | |
FQ214903.1 | - | 4 - 5 | 503 | 69 | ||
FQ215988.1 | - | 5 - 6 | 569 | 69 | ||
FQ216125.1 | - | 4 - 5 | 492 | 69 | ||
FQ216236.1 | - | 2 - 3 | 237 | 69 | ||
FQ216249.1 | - | 5 - 6 | 563 | 69 | ||
FQ216534.1 | - | 4 - 5 | 469 | 69 | ||
FQ217203.1 | - | 4 - 5 | 490 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |