Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sike1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012182.2 | NP_001012182.1 | 4 - 5 | 619 | 90 | |
GenBank mRNA | BC089208.1 | - | 4 - 5 | 619 | 7301503 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |