Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008313.2 | NP_001008314.2 | 5 - 6 | 498 | 65 | |
XM_006236484.2 | XP_006236546.1 | 5 - 6 | 1206 | 65 | ||
XM_006236485.2 | XP_006236547.1 | 5 - 6 | 826 | 65 | ||
GenBank mRNA | BC086595.1 | - | 4 - 5 | 364 | 65 | |
BC086601.1 | - | 5 - 6 | 498 | 7313954 | 65 | |
BC127508.1 | - | 5 - 6 | 513 | 7380822 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |