Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236460.2 | XP_006236522.1 | 1 - 2 | 175 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |