Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191972.1 | NP_001178901.1 | 1 - 2 | 432 | 75 |
Gene Symbol: NEWGENE_1565788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762952.1 | XP_008761174.1 | 1 - 2 | 443 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |