Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ildr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127536.3 | NP_001121008.2 | 3 - 4 | 470 | 73 | |
XM_006248394.2 | XP_006248456.1 | 3 - 4 | 539 | 73 | ||
GenBank mRNA | BC166709.1 | - | 3 - 4 | 470 | 7929589 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |