Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014230.1 | NP_001014252.1 | 3 - 4 | 347 | 80 | |
XM_006248455.2 | XP_006248517.1 | 3 - 4 | 540 | 80 | ||
XM_006248456.2 | XP_006248518.1 | 3 - 4 | 742 | 80 | ||
GenBank mRNA | BC087078.1 | - | 3 - 4 | 347 | 7135432 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |