Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039259.1 | NP_001034348.1 | 5 - 6 | 918 | 78 | |
XM_006248447.2 | XP_006248509.1 | 5 - 6 | 920 | 78 | ||
XM_008768710.1 | XP_008766932.1 | 5 - 6 | 926 | 78 | ||
GenBank mRNA | AY886535.1 | - | 5 - 6 | 918 | 78 | |
BC127441.1 | - | 5 - 6 | 920 | 7111596 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |