Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025015.1 | NP_001020186.1 | 5 - 6 | 803 | 121 | |
XM_006248340.2 | XP_006248402.1 | 4 - 5 | 729 | 121 | ||
GenBank mRNA | BC091236.1 | - | 5 - 6 | 803 | 7322967 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |