Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248111.2 | XP_006248173.1 | 8 - 9 | 2971 | 60 | |
XM_006248112.2 | XP_006248174.1 | 8 - 9 | 3038 | 60 | ||
XM_006248114.2 | XP_006248176.1 | 7 - 8 | 2832 | 60 | ||
XM_008768611.1 | XP_008766833.1 | 8 - 9 | 2170 | 60 | ||
XM_008768612.1 | XP_008766834.1 | 7 - 8 | 2035 | 60 | ||
XM_221630.8 | XP_221630.6 | 7 - 8 | 2151 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |