Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L1td1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001063462.4 | XP_001063462.2 | 1 - 2 | 91 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |