Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107956.1 | NP_001101426.1 | 4 - 5 | 733 | 69 | |
XM_008763676.1 | XP_008761898.1 | 4 - 5 | 1389 | 69 | ||
GenBank mRNA | EF187254.1 | - | 4 - 5 | 735 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |