Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213624.1 | NP_998789.1 | 198 | 94 | ||
GenBank mRNA | AJ699423.1 | - | 198 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus |