Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024261.2 | NP_001019432.1 | 3 - 4 | 435 | 69 | |
XM_008763191.1 | XP_008761413.1 | 3 - 4 | 484 | 69 | ||
XM_008763192.1 | XP_008761414.1 | 3 - 4 | 440 | 69 | ||
XM_008763193.1 | XP_008761415.1 | 3 - 4 | 394 | 69 | ||
GenBank mRNA | BC095841.1 | - | 2 - 3 | 360 | 7387807 | 69 |
BC105612.1 | - | 3 - 4 | 435 | 7135310 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |