Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135085.1 | NP_001128557.1 | 1 - 2 | 109 | 65 | |
GenBank mRNA | BC168750.1 | - | 1 - 2 | 75 | 6921773 | 65 |
BI285268.1 | - | 3 - 4 | 65 | |||
FQ217038.1 | - | 1 - 2 | 75 | 65 | ||
FQ221105.1 | - | 1 - 2 | 107 | 65 | ||
FQ221517.1 | - | 1 - 2 | 75 | 65 | ||
FQ221580.1 | - | 1 - 2 | 136 | 65 | ||
FQ221827.1 | - | 1 - 2 | 77 | 65 | ||
FQ222397.1 | - | 1 - 2 | 75 | 65 | ||
FQ222427.1 | - | 1 - 2 | 78 | 65 | ||
FQ222653.1 | - | 1 - 2 | 124 | 65 | ||
FQ223239.1 | - | 1 - 2 | 75 | 65 | ||
FQ223409.1 | - | 1 - 2 | 124 | 65 | ||
FQ228483.1 | - | 1 - 2 | 78 | 65 | ||
FQ228534.1 | - | 1 - 2 | 75 | 65 | ||
FQ228634.1 | - | 1 - 2 | 83 | 65 | ||
FQ228635.1 | - | 1 - 2 | 120 | 65 |
Gene Symbol: LOC687679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748839.2 | XP_003748887.1 | 1 | 76 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |